Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124933.1 | NP_001118405.1 | 3 - 4 | 656 | 92 | |
NM_001202697.1 | NP_001189626.1 | 3 - 4 | 655 | 92 | ||
NM_128439.3 | NP_180446.1 | 3 - 4 | 972 | 92 | ||
NM_179794.2 | NP_850125.1 | 3 - 4 | 656 | 92 | ||
GenBank mRNA | BT002436.1 | - | 3 - 4 | 661 | 92 | |
BT006602.1 | - | 3 - 4 | 562 | 92 | ||
U89272.1 | - | 3 - 4 | 598 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |