Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338188.1 | NP_001319562.1 | 2 - 3 | 147 | 71 | |
NM_001338189.1 | NP_001326173.1 | 1 - 2 | 283 | 71 | ||
NM_112449.6 | NP_188200.1 | 2 - 3 | 383 | 71 | ||
GenBank mRNA | BT015051.1 | - | 1 - 2 | 208 | 71 | |
BT033124.1 | - | 1 - 2 | 61 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |