Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084699.1 | NP_001078168.1 | 2 - 3 | 670 | 73 | |
NM_001338284.1 | NP_001327579.1 | 3 - 4 | 596 | 73 | ||
NM_112611.3 | NP_188359.2 | 3 - 4 | 892 | 73 | ||
GenBank mRNA | BT025338.1 | - | 2 - 3 | 495 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |