Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338307.1 | NP_001326341.1 | 8 - 9 | 1802 | 73 | |
NM_112651.5 | NP_566585.1 | 8 - 9 | 1900 | 73 | ||
GenBank mRNA | AF148541.1 | - | 8 - 9 | 1779 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |