Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOC64-III | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338317.1 | NP_001325422.1 | 8 - 9 | 1315 | 114 | |
NM_112678.4 | NP_188424.2 | 8 - 9 | 1360 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |