Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112791.6 | NP_188535.4 | 16 - 17 | 2387 | 91 | |
GenBank mRNA | DQ399530.1 | - | 15 - 16 | 2187 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |