Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP47B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112800.4 | NP_188544.1 | 4 - 5 | 1383 | 75 | |
GenBank mRNA | AK226472.1 | - | 4 - 5 | 1324 | 75 | |
BT026489.1 | - | 4 - 5 | 1099 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |