Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP81H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119895.4 | NP_568024.2 | 2 - 3 | 1168 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |