Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085042.1 | NP_001078511.1 | 3 - 4 | 1166 | 85 | |
NM_119974.4 | NP_195526.1 | 3 - 4 | 1184 | 85 | ||
GenBank mRNA | AK226389.1 | - | 3 - 4 | 1158 | 85 | |
AY093153.1 | - | 3 - 4 | 1134 | 85 | ||
BT008873.1 | - | 2 - 3 | 973 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |