Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP60-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202736.1 | NP_001189665.1 | 1 - 2 | 212 | 64 | |
NM_179872.3 | NP_850203.1 | 1 - 2 | 240 | 64 | ||
GenBank mRNA | AK175415.1 | - | 1 - 2 | 199 | 64 | |
AY136360.1 | - | 1 - 2 | 76 | 64 | ||
BT000103.1 | - | 1 - 2 | 36 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |