Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100028.3 | NP_171653.2 | 5 - 6 | 931 | 76 | |
GenBank mRNA | BT028973.1 | - | 4 - 5 | 610 | 76 | |
DQ056443.1 | - | 4 - 5 | 610 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |