Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100097.5 | NP_171719.2 | 4 - 5 | 1264 | 102 | |
GenBank mRNA | AY219826.1 | - | 4 - 5 | 1045 | 102 | |
AY322525.1 | - | 4 - 5 | 1045 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |