Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100144.3 | NP_563659.1 | 2 - 3 | 960 | 65 | |
GenBank mRNA | AF367266.1 | - | 2 - 3 | 773 | 65 | |
AY143952.1 | - | 2 - 3 | 685 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |