Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100351.5 | NP_171965.1 | 6 - 7 | 579 | 89 | |
GenBank mRNA | AY099741.1 | - | 6 - 7 | 555 | 89 | |
BT003415.1 | - | 6 - 7 | 523 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |