Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331495.1 | NP_001322020.1 | 12 - 13 | 1736 | 88 | |
NM_001331496.1 | NP_001322021.1 | 10 - 11 | 1208 | 88 | ||
NM_001331497.1 | NP_001322022.1 | 12 - 13 | 1759 | 88 | ||
NM_100352.3 | NP_171966.2 | 12 - 13 | 1776 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |