Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331495.1 | NP_001322020.1 | 8 - 9 | 1475 | 116 | |
NM_001331496.1 | NP_001322021.1 | 6 - 7 | 947 | 116 | ||
NM_001331497.1 | NP_001322022.1 | 8 - 9 | 1498 | 116 | ||
NM_100352.3 | NP_171966.2 | 8 - 9 | 1515 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |