Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP88A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331521.1 | NP_001320198.1 | 3 - 4 | 1047 | 72 | |
NM_100394.4 | NP_172008.1 | 3 - 4 | 1114 | 72 | ||
GenBank mRNA | AK229009.1 | - | 3 - 4 | 1049 | 72 | |
BT029169.1 | - | 2 - 3 | 713 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |