Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197984.1 | NP_001184913.1 | 28 - 29 | 3038 | 120 | |
NM_001331555.1 | NP_001322482.1 | 27 - 28 | 3252 | 120 | ||
NM_001331556.1 | NP_001322483.1 | 26 - 27 | 3086 | 120 | ||
NM_100436.5 | NP_563743.2 | 27 - 28 | 3335 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |