Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331560.1 | NP_001322344.1 | 3 - 4 | 1006 | 75 | |
NM_100441.3 | NP_172052.2 | 3 - 4 | 1026 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |