Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100511.3 | NP_172119.1 | 10 - 11 | 1789 | 79 | |
GenBank mRNA | AF253474.1 | - | 10 - 11 | 1627 | 79 | |
AK176257.1 | - | 10 - 11 | 1645 | 79 | ||
AY099579.1 | - | 10 - 11 | 1645 | 79 | ||
BT008413.1 | - | 10 - 11 | 1574 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |