Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftsh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100625.4 | NP_172231.2 | 7 - 8 | 3219 | 126 | |
GenBank mRNA | AY064138.1 | - | 7 - 8 | 2277 | 126 | |
AY124808.1 | - | 7 - 8 | 1965 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |