Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112965.1 | NP_188710.1 | 10 - 11 | 1118 | 116 | |
GenBank mRNA | AF129516.1 | - | 10 - 11 | 1118 | 116 | |
AK117114.1 | - | 10 - 11 | 975 | 116 | ||
BT005326.1 | - | 10 - 11 | 920 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |