Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337108.1 | NP_001323852.1 | 7 - 8 | 1422 | 84 | |
NM_130060.4 | NP_182022.2 | 7 - 8 | 1428 | 84 | ||
GenBank mRNA | AY081322.1 | - | 7 - 8 | 1367 | 84 | |
BT010360.1 | - | 7 - 8 | 1023 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |