Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337197.1 | NP_001325285.1 | 6 - 7 | 1399 | 116 | |
NM_001337198.1 | NP_001325284.1 | 5 - 6 | 1426 | 116 | ||
NM_001337199.1 | NP_001325283.1 | 4 - 5 | 1391 | 116 | ||
NM_130212.3 | NP_182171.1 | 5 - 6 | 1316 | 116 | ||
GenBank mRNA | KJ676986.1 | - | 4 - 5 | 1391 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |