Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130215.3 | NP_566077.1 | 1 | 896 | 138 | |
GenBank mRNA | AY052323.1 | - | 1 | 655 | 138 | |
AY084510.1 | - | 1 | 669 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Both primers and probe map within a single exon |