Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNIFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100722.5 | NP_172325.2 | 5 - 6 | 1007 | 122 | |
GenBank mRNA | AF419347.1 | - | 5 - 6 | 896 | 122 | |
AY078068.1 | - | 5 - 6 | 896 | 122 | ||
AY094425.1 | - | 5 - 6 | 938 | 122 | ||
AY149950.1 | - | 5 - 6 | 896 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |