Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125184.3 | NP_568873.1 | 1 - 2 | 491 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |