Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftsh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125277.4 | NP_568892.1 | 5 - 6 | 1543 | 97 | |
GenBank mRNA | AY059856.1 | - | 5 - 6 | 1350 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |