Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203645.2 | NP_001190574.1 | 9 - 10 | 1823 | 73 | |
NM_001203646.1 | NP_001190575.1 | 8 - 9 | 1860 | 73 | ||
NM_001345386.1 | NP_001331790.1 | 7 - 8 | 2375 | 73 | ||
NM_001345387.1 | NP_001331791.1 | 8 - 9 | 2296 | 73 | ||
NM_001345388.1 | NP_001331792.1 | 8 - 9 | 1898 | 73 | ||
NM_001345389.1 | NP_001331789.1 | 8 - 9 | 1851 | 73 | ||
NM_125403.3 | NP_568919.1 | 9 - 10 | 1820 | 73 | ||
GenBank mRNA | AB046956.1 | - | 8 - 9 | 1383 | 73 | |
AB493801.1 | - | 8 - 9 | 1383 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |