Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABG3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036084.2 | NP_001031161.1 | 6 - 7 | 671 | 64 | |
NM_103820.4 | NP_175355.1 | 6 - 7 | 716 | 64 | ||
GenBank mRNA | AB071849.1 | - | 6 - 7 | 553 | 64 | |
AK227548.1 | - | 6 - 7 | 642 | 64 | ||
BT004594.1 | - | 6 - 7 | 553 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |