Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333424.1 | NP_001323380.1 | 2 - 3 | 385 | 80 | |
NM_001333425.1 | NP_001323379.1 | 1 - 2 | 477 | 80 | ||
NM_103902.4 | NP_564562.1 | 2 - 3 | 395 | 80 | ||
GenBank mRNA | BT004060.1 | - | 2 - 3 | 322 | 80 | |
BT005145.1 | - | 2 - 3 | 306 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |