Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085090.2 | NP_001078559.1 | 5 - 6 | 1172 | 147 | |
NM_001343068.1 | NP_001318519.1 | 5 - 6 | 1214 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |