Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NADP-ME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121205.4 | NP_196728.1 | 1 - 2 | 507 | 78 | |
GenBank mRNA | AF428407.1 | - | 1 - 2 | 245 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |