Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128190.3 | NP_180201.1 | 17 - 18 | 2059 | 87 | |
GenBank mRNA | AY035110.1 | - | 17 - 18 | 1368 | 87 | |
FJ708701.1 | - | 17 - 18 | 1292 | 87 | ||
U47029.1 | - | 17 - 18 | 1342 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |