Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336059.1 | NP_001323820.1 | 8 - 9 | 959 | 102 | |
NM_001336060.1 | NP_001323819.1 | 7 - 8 | 1034 | 102 | ||
NM_001336061.1 | NP_001323818.1 | 8 - 9 | 993 | 102 | ||
NM_001336062.1 | NP_001323817.1 | 8 - 9 | 959 | 102 | ||
NM_128192.2 | NP_565621.1 | 8 - 9 | 959 | 102 | ||
GenBank mRNA | AF119572.1 | - | 8 - 9 | 923 | 102 | |
AK117879.1 | - | 8 - 9 | 959 | 102 | ||
BT005340.1 | - | 8 - 9 | 867 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |