Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336097.1 | NP_001323455.1 | 5 - 6 | 706 | 150 | |
NM_128260.5 | NP_180270.1 | 5 - 6 | 552 | 150 | ||
GenBank mRNA | AF043528.1 | - | 5 - 6 | 429 | 150 | |
AF375455.1 | - | 5 - 6 | 419 | 150 | ||
AY124806.1 | - | 5 - 6 | 386 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |