Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT2G28390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128399.4 | NP_029426.1 | 4 - 5 | 991 | 66 | |
GenBank mRNA | AK118037.1 | - | 4 - 5 | 778 | 66 | |
AK175575.1 | - | 4 - 5 | 778 | 66 | ||
AK175699.1 | - | 4 - 5 | 778 | 66 | ||
AK176064.1 | - | 4 - 5 | 788 | 66 | ||
AK176154.1 | - | 4 - 5 | 788 | 66 | ||
AK221211.1 | - | 4 - 5 | 778 | 66 | ||
AY056801.1 | - | 4 - 5 | 771 | 66 | ||
AY133524.1 | - | 4 - 5 | 701 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |