Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128548.4 | NP_180555.2 | 18 - 19 | 3519 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |