Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KINESIN-12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035673.2 | NP_001030750.1 | 4 - 5 | 764 | 62 | |
NM_113271.4 | NP_189009.2 | 4 - 5 | 794 | 62 | ||
GenBank mRNA | AF540489.1 | - | 4 - 5 | 766 | 62 | |
AY124006.1 | - | 4 - 5 | 762 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |