Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113321.5 | NP_189058.1 | 2 - 3 | 315 | 139 | |
GenBank mRNA | AF104329.1 | - | 2 - 3 | 179 | 139 | |
AY053413.1 | - | 2 - 3 | 195 | 139 | ||
AY085022.1 | - | 2 - 3 | 215 | 139 | ||
AY133587.1 | - | 2 - 3 | 130 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |