Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113392.3 | NP_189124.1 | 4 - 5 | 525 | 72 | |
GenBank mRNA | AJ224307.1 | - | 4 - 5 | 502 | 72 | |
AY080799.1 | - | 4 - 5 | 523 | 72 | ||
AY114077.1 | - | 4 - 5 | 381 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |