Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085321.2 | NP_001078790.1 | 1 - 2 | 408 | 66 | |
NM_125748.3 | NP_568974.1 | 2 - 3 | 685 | 66 | ||
GenBank mRNA | AF360327.1 | - | 2 - 3 | 332 | 66 | |
AY056337.1 | - | 1 - 2 | 296 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |