Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126014.1 | NP_001119486.1 | 6 - 7 | 1691 | 124 | |
NM_125749.5 | NP_201159.2 | 5 - 6 | 1625 | 124 | ||
GenBank mRNA | AY735746.1 | - | 5 - 6 | 1590 | 124 | |
AY954880.1 | - | 5 - 6 | 1527 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |