Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125780.3 | NP_201190.1 | 2 - 3 | 480 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |