Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI-4KBETA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125803.4 | NP_201212.1 | 10 - 11 | 3428 | 101 | |
GenBank mRNA | AF462854.1 | - | 10 - 11 | 3396 | 101 | |
AY139795.1 | - | 10 - 11 | 2852 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |