Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125840.4 | NP_201249.2 | 20 - 21 | 1965 | 129 | |
GenBank mRNA | AY308736.1 | - | 20 - 21 | 1881 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |