Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101047.3 | NP_563893.1 | 2 - 3 | 284 | 147 | |
GenBank mRNA | AF348580.1 | - | 2 - 3 | 151 | 147 | |
AK229049.1 | - | 2 - 3 | 213 | 147 | ||
AY087248.1 | - | 2 - 3 | 282 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |