Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH10A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198470.1 | NP_001185399.1 | 5 - 6 | 727 | 70 | |
NM_106150.4 | NP_565094.1 | 5 - 6 | 835 | 70 | ||
GenBank mRNA | AY087395.1 | - | 5 - 6 | 742 | 70 | |
AY093071.1 | - | 5 - 6 | 700 | 70 | ||
BT008872.1 | - | 5 - 6 | 571 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |