Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334710.1 | NP_001323338.1 | 11 - 12 | 1402 | 67 | |
NM_106262.4 | NP_177740.1 | 11 - 12 | 1037 | 67 | ||
GenBank mRNA | AK221564.1 | - | 11 - 12 | 1037 | 67 | |
AY084871.1 | - | 11 - 12 | 1033 | 67 | ||
BT025560.1 | - | 10 - 11 | 950 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |