Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084367.2 | NP_001077836.1 | 6 - 7 | 3188 | 84 | |
NM_106379.9 | NP_177854.6 | 6 - 7 | 3340 | 84 | ||
GenBank mRNA | EU014690.1 | - | 5 - 6 | 3053 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |